Sodium dihydrogen phosphate dihydrate 98.0-102.0%, BAKER ANALYZED®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate

Management of Change (MOC) category = NI

BAKR3819-01 BAKR3819-05
CAJT3819-1EA 361.58 CAD
CAJT3819-1 CAJT3819-5
Sodium dihydrogen phosphate dihydrate 98.0-102.0%, BAKER ANALYZED®
Sodium dihydrogen phosphate dihydrate

Lot analysis on label.

Formula: NaH₂PO₄·2H₂O
Boiling Pt: ? 650 °C (1013 hPa)
Melting Pt: 60 °C
Density: 1.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149209
CAS Number: 13472-35-0
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (NaH₂PO₄·2H₂O) 98.0 - 102.0 %
pH of 5% Solution at 25°C 4.1 - 4.5
Chloride (Cl) ≤5 ppm
Insoluble Matter, Calcium and NH₄OH Precipitate ≤0.01 %
Nitrogen Compounds (as N) ≤0.001 %
ACS - Sulfate (SO₄) ≤0.003 %
Arsenic (As) ≤0.5 ppm
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR