Sodium molybdate(VI) dihydrate 99.5-103.0%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate

Management of Change (MOC) category = N

BAKR3764-01
CAJT3764-1EA 869.71 CAD
CAJT3764-1 CAJT3764-5 CAJT3764-07 CAJT3764-R
Sodium molybdate(VI) dihydrate 99.5-103.0%, BAKER ANALYZED® ACS, J.T.Baker®
Sodium molybdate(VI) dihydrate

Lot analysis on label.

Formula: Na₂MoO₄·2H₂O
MW: 241.95 g/mol
Melting Pt: 687
Density: 3.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149170
CAS Number: 10102-40-6
EINECS: 231-551-7
Merck Index: 13,08718

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (Na₂MoO₄·2H₂O) 99.5 - 103.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 7.0 - 10.5
Chloride (Cl) ≤0.005 %
Ammonium (NH₄) ≤0.001 %
Sulfate (SO₄) ≤0.015 %
Trace Impurities - Iron (Fe) ≤0.001 %
Phosphate (PO₄) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR