Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

BAKR3717-01 BAKR3717-07
CAJT3717-01EA 226.97 CAD
CAJT3717-01 CAJT3717-07
Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Sodium hydroxide
Formula: NaOH
Boiling Pt: 1390 °C
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
Meets ChP Chemical Specifications
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets B.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification A (Sodium) Passes Test
NF - Identification B (pH) ≥11.0
NF - Insoluble Substances and Organic Matter Passes Test
NF - Potassium (K) ≤0.5 %
NF - Carbonate (as Na₂CO₃) ≤3.0 %
NF - Total Alkali 95.0 - 100.5 %
NF - Content of Sodium 54.0 - 59.8 %
ChP -Assay 97.0 - 100.5 %
ChP -Sodium carbonate 2.0% max
ChP -Identification Passes Test
ChP -Alkalinity Passes Test
ChP -Clarity and color of solution  Passes Test
ChP -Chloride  0.005 % max
ChP -Sulfate  0.015 % max
ChP -Potassium  Passes Test
ChP -Aluminum and iron   0.1% max
ChP -Iron  0.001% max
ChP -Heavy metal  20 ppm max
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Carbonate (as Na₂CO₃) ≤2.0 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Sulfate (SO₄) ≤200 ppm
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Assay (NaOH) 97.0 - 100.5 %
JP - Assay 95.0 - 101.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.050 %
JP - Heavy Metals (as Pb) ≤30 ppm
JP - Potassium (K) Passes Test
JP - Carbonate (as Na₂CO₃) ≤2.0 %
JP - Mercury (Hg) Passes Test
Endotoxin Concentration (EU/g) ≤5
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR