Sodium carbonate monohydrate 99.5-100.5% (by anhydrous basis), crystals NF, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = R

BAKR3603-07 BAKR3603-09
CAJT3603-7EA 2920.71 CAD
CAJT3603-7 CAJT3603-09
Sodium carbonate monohydrate 99.5-100.5% (by anhydrous basis), crystals NF, Multi-Compendial, J.T.Baker®
Sodium carbonate monohydrate
Formula: Na₂CO₃·1H₂O
Melting Pt: 107.85 °C
Density: 2.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149177
CAS Number: 5968-11-6
EINECS: 207-838-8

Order Now

Specification Test Results

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Water (H₂O) 12.0 - 15.0 %
NF - Assay (Na₂CO₃) (anhydrous basis) 99.5 - 100.5 %
NF - Endotoxin Concentration (EU/g) ≤ 2.5
EP/BP - Assay (Na₂CO₃) 83.0 - 87.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Alkali Hydroxides and Bicarbonates Passes Test
EP/BP - Chloride (Cl) ≤ 125 ppm
EP/BP - Sulfate (SO₄) ≤ 250 ppm
EP/BP - Iron (Fe) ≤ 50 ppm
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR