Potassium thiocyanate ≥99% (Ag basis), crystals, BAKER ANANYZED™ ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Potassium rhodanide

Management of Change (MOC) category = NI

BAKR3326-R
CAJT3326-REA 27029.93 CAD
CAJT3326-R
Potassium thiocyanate ≥99% (Ag basis), crystals, BAKER ANANYZED™ ACS
Potassium thiocyanate

Crystals. Lot analysis on label.

Formula: KSCN
Boiling Pt: 500 °C (1013 hPa)
Melting Pt: 175 °C
Density: 1.61 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (KSCN) (by Ag titrn) ≥98.5 %
Insoluble in H₂O ≤0.005 %
pH of 5% Solution at 25°C 5.3 - 8.7
Chloride (Cl) ≤0.005 %
Sulfate (SO₄) ≤0.005 %
Ammonium (NH₄) ≤0.003 %
Sodium (Na) ≤0.005 %
Iodine Consuming Substances (meq/g) ≤0.004
Use Test Passes Test
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR