Potassium hydroxide ≥85.0%, pellets, BAKER ANALYZED®, Electronic Grade, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Potash caustic, Caustic potash

Management of Change (MOC) category = R

3141-07 BAKR3141-01 BAKR3141-05 BAKR3141-19
JT3141-7EA 781.53 CAD
JT3141-7 CAJT3141-1 CAJT3141-5 CAJT3141-19
Potassium hydroxide ≥85.0%, pellets, BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Potassium hydroxide

Pellets.

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

For Microelectronic Use
Assay (KOH) ≥85.0 %
Potassium Carbonate (K₂CO₃) ≤1.0 %
Insoluble Matter ≤50 ppm
Chloride (Cl) ≤0.01 %
Sulfate (SO₄) ≤20 ppm
Ammonium Hydroxide Precipitate ≤0.02 %
Sodium (Na) ≤0.05 %
Nitrogen Compounds (as N) ≤10 ppm
Phosphate (PO₄) ≤5 ppm
Heavy Metals (as Ag) ≤10 ppm
Iron (Fe) ≤10 ppm
Mercury (Hg), For Information Only
Nickel (Ni) ≤10 ppm
Calcium (Ca) ≤50 ppm
Magnesium (Mg), For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR