Potassium hexacyanoferrate(III) ≥99.0% (by iodometry), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide

Management of Change (MOC) category = N

BAKR3104-07 BAKR3104-01 BAKR3104-05
CAJT3104-7EA 5879.37 CAD
CAJT3104-7 CAJT3104-1 CAJT3104-5
Potassium hexacyanoferrate(III) ≥99.0% (by iodometry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium hexacyanoferrate (III)

  • Lot analysis on label

Formula: K₃Fe(CN)₆
MW: 329.25 g/mol
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3
Merck Index: 13,07712

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (K₃Fe(CN)₆) (by iodometry) ≥99.0 %
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.01 %
Sulfate (SO₄) ≤0.01 %
Ferro Compounds (as [Fe(CN)₆]⁴⁻) ≤0.05 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR