Potassium carbonate sesquihydrate 98.5-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

BAKR3010-R BAKR3010-01 BAKR3010-05
CAJT3010-REA 17703.24 CAD
CAJT3010-R CAJT3010-1 CAJT3010-5
Potassium carbonate sesquihydrate 98.5-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium carbonate sesquihydrate
Formula: CK₂O₃·1,5H₂O
Melting Pt: 891 °C
Density: 2.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149925
CAS Number: 6381-79-9
EINECS: 209-529-3

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (K₂CO₃·1.5H₂O) 98.5 - 101.0 %
ACS - Insoluble Matter ≤0.01 %
ACS - Loss on Heating at 285°C 14.0 - 16.5 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Chloride (Cl) ≤0.003 %
ACS - Magnesium (Mg) ≤0.002 %
ACS - Nitrogen Compounds (as N) ≤0.001 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Silica (SiO₂) ≤0.005 %
ACS - Sulfur Compounds (as SO₄) ≤0.004 %
ACS - Ammonium Hydroxide Precipitate ≤0.01 %
ACS - Sodium (Na) ≤0.02 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR