Potassium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

BAKR2997-01 BAKR2997-07
CAJT2997-1CS 3622.61 CAD
CAJT2997-1 CAJT2997-7
Potassium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium bromide
Formula: KBr
Boiling Pt: 1435 °C (1013 hPa)
Melting Pt: 730 °C
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (KBr) (by Ag titrn) ≥99.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 5.0 - 8.8
Bromate (BrO₃) ≤0.001 %
Iodate (IO₃) ≤0.001 %
Chloride (Cl) ≤0.2 %
Iodide (I) ≤0.001 %
Nitrogen Compounds (as N) ≤0.005 %
Sulfate (SO₄) ≤0.005 %
Barium (Ba) ≤0.002 %
Calcium (Ca) ≤0.002 %
Magnesium (Mg) ≤0.001 %
Sodium (Na) ≤0.02 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR