Molecular sieve 3A (0.3 nm, 3 Å), BAKER ANALYZED® 8-12 Mesh, activated, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate

Management of Change (MOC) category = N

BAKR2710-05
CAJT2710-1EA 759.59 CAD
CAJT2710-1 CAJT2710-5
Molecular sieve 3A (0.3 nm, 3 Å), BAKER ANALYZED® 8-12 Mesh, activated, J.T.Baker®
Molecular sieve 3A (0.3 nm, 3 Å)

Pellets.

Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number: MFCD00131613
CAS Number: 308080-99-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Loss on Drying at 150°C ≤ 5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR