Calcium oxide, BAKER ANANYZED™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Quicklime, Calcium oxide from marble, caustic, Lime

M.O.C. Category N

BAKR1410-07 BAKR1410-01
CAJT1410-7EA 1089.36 CAD
CAJT1410-7 CAJT1410-1 CAJT1410-5
Calcium oxide, BAKER ANANYZED™
Calcium oxide

Powder. Lot analysis on label.

Formula: CaO
MW: 56.08 g/mol
Boiling Pt: 2850
Melting Pt: 2580
Density: 3.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010911
CAS Number: 1305-78-8
EINECS: 215-138-9
Merck Index: 13,01691

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Insoluble in CH₃COOH and NH₄OH Precipitate ≤1.0 %
Loss on Ignition ≤5.0 %
Chloride (Cl) ≤0.005 %
Nitrate (NO₃) ≤0.05 %
Sulfate (SO₄) ≤0.1 %
Heavy Metals (as Pb) ≤0.005 %
Iron (Fe) ≤0.1 %
Assay, For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR