Ammonium sulfate 99.0-100.5%, granules ACS, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = RL

BAKR0795-08 BAKR0795-12 BAKR0795-65
CAJT0795-8EA 7866.09 CAD
CAJT0795-8 CAJT0795-12 CAJT0795-65
Ammonium sulfate 99.0-100.5%, granules ACS, FCC, J.T.Baker®
Ammonium sulphate

Food GMP manufactured product.

Formula: (NH₄)₂SO₄
Melting Pt: 336…339 °C
Density: 1.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
ACS - Assay ((NH₄)₂SO₄) 99.0 - 100.5 %
ACS - Insoluble Matter ≤0.005 %
ACS - Residue on Ignition (ACS) ≤0.005 %
ACS - pH of 5% Solution at 25°C 5.0 - 6.0
ACS - Nitrate (NO₃) ≤0.001 %
Chloride (Cl) ≤3 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Phosphate (PO₄) ≤3 ppm
Heavy Metals (as Pb) ≤3 ppm
FCC - Assay ((NH₄)₂SO₄)(F.C.C.) 99.0 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Residue on Ignition (FCC) ≤0.25 %
FCC - Lead (Pb) ≤3 mg/kg
FCC - Selenium (Se) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR