Ammonium sulfate ≥99%, granules, BAKER ANANYZED™ ACS, FCC for biochemistry, for chromatography endotoxin tested, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

Management of Change (MOC) category = RL

BAKR0793-07 BAKR0793-08 BAKR0793-09
CAJT0793-7EA 2233.27 CAD
CAJT0793-7 CAJT0793-8 CAJT0793-9
Ammonium sulfate ≥99%, granules, BAKER ANANYZED™ ACS, FCC for biochemistry, for chromatography endotoxin tested, J.T.Baker®
Ammonium sulphate

Ideal for biotechnology, cell biology and molecular biology discovery, and biopharmaceutical production, J.T.Baker® brand denaturants offer high purity, low insolubles and heavy metals with minimal contaminants. Extensive experience in managing the risks associated with handling and mixing large volumes of corrosive materials has enabled us to provide custom solutions perfectly suited to your needs.


  • Food GMP manufactured product

Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: 336…339 °C
Density: 1.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets ACS Reagent Chemical Requirements
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
ACS - Assay ((NH₄)₂SO₄) 99.0 - 100.5 %
ACS - Insoluble Matter ≤0.005 %
ACS - Residue on Ignition (ACS) ≤0.005 %
ACS - pH of 5% Solution at 25°C 5.0 - 6.0
ACS - Nitrate (NO₃) ≤0.001 %
Chloride (Cl) ≤3 ppm
Phosphate (PO₄) ≤3 ppm
Heavy Metals (as Pb) ≤3 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
FCC - Assay ((NH₄)₂SO₄) 99.0 - 100.5 %
FCC - Identification Passes Test
FCC - Residue on Ignition (FCC) ≤0.25 %
FCC - Lead (Pb) ≤3 mg/kg
FCC - Selenium (Se) ≤0.003 %
Endotoxin Concentration (2.5 EU/g max.) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR