Ammonium sulfate 38% (w/w) in aqueous solution, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

Management of Change (MOC) category = N

BAKR0791-09
CAJT0791-9EA 8155.12 CAD
CAJT0791-9
Ammonium sulfate 38% (w/w) in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Density: 1.77 g/cm³
MDL Number: MFCD00003391
CAS Number: 7783-20-2
Merck Index: 13,00559

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay ((NH₄)₂SO₄) (by formol method) 37.0 - 39.0 %
Color (APHA) ≤10
pH (As Is" basis)" 5.7 - 6.3
Identification A Passes Test
Identification B Passes Test
Insoluble Matter ≤0.005 %
Residue after Ignition ≤0.005 %
Nitrate (NO₃) ≤0.001 %
Chloride (Cl) ≤0.0005 %
Phosphate (PO₄) ≤0.0005 %
Arsenic (As) ≤0.00002 %
Heavy Metals (as Pb) ≤0.0005 %
Iron (Fe) ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR