Silicic acid hydrate (≥84% SiO₂), powder, BAKER ANALYZED®, J.T.Baker®

Supplier: VWR International


Management of Change (MOC) category = N

CAJT0324-5 BAKR0324-01
CAJT0324-5EA 2743.07 CAD
CAJT0324-5 CAJT0324-1
Silicic acid hydrate (≥84% SiO₂), powder, BAKER ANALYZED®, J.T.Baker®
Silicic acid hydrate

Powder.

Formula: SiO₂·xH₂O
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Density: 2.10 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00054122
CAS Number: 1343-98-2
REACH: 01-2119379499-16

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (as SiO₂) ≥84 %
Nonvolatile with HF ≤0.4 %
Chloride (Cl) ≤0.01 %
Sulfate (SO₄) ≤0.02 %
Heavy Metals (as Pb) ≤0.002 %
Iron (Fe) ≤0.007 %
Loss on Ignition (as H₂O) ≤16 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR