Hydrogen peroxide solution 30% (w/w) stabilized, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

HX0635-2 HX0635-3 HX0635-6
CAHX0635-2CS 1036 CAD
CAHX0635-2 CAHX0635-3 CAHX0635-6
Hydrogen peroxide solution 30% (w/w) stabilized, GR ACS, Supelco®
Hydrogen peroxide

29.0-32.0% W/W

Formula: H₂O₂
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1,II

Order Now

Specification Test Results

Expiration date Exp 24 months from mfg date
Assay 29.0-32.0%
Ammonium (NH4) 5ppm max
Chloride (Cl) 3ppm max
Color (APHA) 10 max
Form Clear liquid
Heavy metals (as Pb) 1ppm max
Iron (Fe) 0.5ppm max
Nitrate (NO3) 2ppm max
Phosphate (PO4) 2ppm max
Residue after evaporation 0.002% max
Sulfate (SO4) 5ppm max
Titrable acid 0.0006meq/g max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR