Hexane (mixture of isomers) 98,5 ACS HPLC, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hexanes, Isohexane

HX0290-20 HX0290-6 HX0290-NP20 HX0290-1
CAHX0290-20EA 6438 CAD
CAHX0290-20 CAHX0290-6 CAHX0290-NP20 CAHX0290-1
Hexane (mixture of isomers) 98,5 ACS HPLC, Supelco®
Hexane (mixture of isomers)

HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: -95
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (n-hexane) Report
Assay (total hexane isomers + methylcyclopentane) 98.5% min
Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Infrared spectrum Conforms
Residue after evaporation 5ppm max
Sulfur compounds (as S) 0.005% max
Thiophene To pass test
UV Abs. at 200 nm 1.00AU max
UV Abs. at 210 nm 0.30AU max
UV Abs. at 220 nm 0.20AU max
UV Abs. at 230 nm 0.10AU max
UV Abs. at 240 nm 0.04AU max
UV Abs. at 250 nm 0.02AU max
UV Abs. at 270 nm 0.01AU max
UV Abs. at 300 nm 0.01AU max
UV Abs. at 350 nm 0.01AU max
UV Cut-off 200nm max
Water (H2O) 0.01% max
Water-soluble titrable acid 0.0003meq/g max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR