Ultragrade 19 Vacuum Pump Oil, Edwards
Supplier: Edwards
H11025015
CAH110-25-015EA 115.55 CAD
CAH110-25-015
Ultragrade 19 Vacuum Pump Oil, Edwards
Lubricants
This oil has been produced by a unique hydrotreating process
When combined with additives, the Ultragrade® oils give improved vacuum performance, thermal and oxidation stability and rust inhibition. The anti-oxidants included in the oils enable the pump to withstand high operating temperatures without oil degradation, so extending intervals between oil changes.
This process eliminates nitrogen, sulphur and oxygen and the aromatic hydrocarbons, leaving a clear base stock essentially free of impurities.
Pour Point: -16°