CrossLab Torches for 5100/5110 and 5800/5900 ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G8010-60228 G8010-60231 G8010-60232 G8010-60236 G8020-68002 G8010-60237 G8020-68005 G8010-60234 G8012-60000 G8020-68001 G8020-68003 G8020-68004 G8020-68007
CAG8010-60228EA 1715.57 CAD
CAG8010-60228 CAG8010-60231 CAG8010-60232 CAG8010-60236 CAG8020-68002 CAAGG801060237 CAAGG802068005 CAAGG801060234 CAAGG801260000 CAAGG802068001 CAAGG802068003 CAAGG802068004 CAAGG802068007
CrossLab Torches for 5100/5110 and 5800/5900 ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
One piece, semi and fully demountable RV/DV torches in a variety of injector sizes and materials.
  • Fully demountable torch provides the flexibility of having a variety of injector sizes and materials to handle difficult and diverse application demands
  • Ease of maintenence with a tool-ree removable injector, all with no optimization required
  • New organics outer tube set is made of high purity quartz providing longer lifetime when using organic solvents
  • The vertical torch orientation provides the ultimate configuration for determining tough samples
  • Handles even
More Product Information