Nebulizer Supplies for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3266-80015 G3266-80020 G3266-80012 PL0860-0550
CAG3266-80015EA 75.75 CAD
CAG3266-80015 CAG3266-80020 CAAGG326680012 AGPL0860-0550
Nebulizer Supplies for ICP-OES
Chromatography Supplies Chromatography Spare Parts
Gas/sample line connectors, tubing, and cleaning tools to fit a variety of ICP-OES nebulizers.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
The Agilent ICP-OES series is compatible with a range of accessories and supplies designed to extend the capabilities of the instruments. Agilent has a variety of nebulizer gas/sample connectors, capillary tubing, and cleaningMore Product Information