Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
EX0995-1
CAEX0995-1CS 738 CAD
CAEX0995-1 CAEX0995-5
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
Cleaning Agents Detergents and Surface Cleansers
When diluted with water, concentrates can clean by themselves without leaving residues
Extran MN 01 Powdered Detergent is a multi-purpose detergent for heavy-duty laboratory cleaning applications. This mixture of anionic surfactants and auxiliary agents can be used on glass, plastic, rubber, metal, and quartz. Phosphate-free. Items to be cleaned should be immersed in a cleaning solution and thoroughly brushed or wiped. Dose 2 to 5 g/L. pH of 0.2% solution is 10.9.
For routine laboratory applications, no scrubbing is necessary once items are removed from the cleaning bath. For bestMore Product Information