Ethyl acetate ≥99.9% (by GC), OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

EX0241-NP20 EX0241-1 EX0241-6 EX0241P-1
CAEX0241-NP20EA 1532 CAD
CAEX0241-NP20 CAEX0241-1 CAEX0241-6 CAEX0241P-1
Ethyl acetate ≥99.9% (by GC), OmniSolv®, Supelco®
Ethyl acetate

For spectrophotometry, chromatography, and residue analysis. UV cutoff 254nm. Certificate of Analysis on label.

Formula: CH₃COOC₂H₅
Boiling Pt: 76.5…77.5 °C (lit.)
Melting Pt: –84 °C (lit.)
Density: 0.902 g/ml (25°C) (lit.)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Capillary ECD responsive substances (as PCNB) 2ng/L max
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 250ppt max
Form Clear liquid
Identity (IR-spectrum) Conforms
Residue after evaporation 1ppm max
Titrable acid 0.8µeq/g max
UV Abs. at 254 nm 1.00AU max
UV Abs. at 255 nm 0.80AU max
UV Abs. at 257 nm 0.35AU max
UV Abs. at 265 nm 0.02AU max
UV Abs. at 270 nm 0.01AU max
UV Abs. at 300 nm 0.005AU max
UV Cut-off 254nm max
Water (H2O) 0.02% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR