Dichloromethane ≥99.9% stabilised, OmniSolv® HR-GC for gas chromatography, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Methylene chloride, DCM

DX0837-1
CADX0837-1CS 1685 CAD
CADX0837-1
Dichloromethane ≥99.9% stabilised, OmniSolv® HR-GC for gas chromatography, Supelco®
Dichloromethane

For high resolution gas chromatography. Lot analysis and chromatograms on label.

Formula: CH₂Cl₂
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC) 99.9% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR