Chloroform ≥99% stabilized, Supelco®

Supplier: MilliporeSigma
Danger

CX1060-7 CX1060-1
CACX1060-7CS 1810 CAD
CACX1060-7 CA-CX1060-1
Chloroform ≥99% stabilized, Supelco®
Chloroform

Stabilized with 1% ethanol.

Formula: CHCl₃
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99% min
Acid and phosgene To pass test
Aldehyde and ketone To pass test
Chloride and free chlorine To pass test
Description To pass test
Identity (IR-spectrum) Conforms
Nonvolatile residue 0.002% max
Solubility To pass test
Specific Gravity at 25/25°C 1.475-1.480


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR