Vented Screw Caps for Falcon® Flasks, Sterile, Corning
Supplier: Corning
354637 354638 354639
CACB354637CS 129.91 CAD
CACB354637 CACB354638 CACB354639
Vented Screw Caps for Falcon® Flasks, Sterile, Corning
Closures Bottle Closures
PE with integral hydophobic 0.2 µm microporous membrane filter vent.
Vented caps minimize contamination associated with standard open incubation. Gases required for cell growth and metabolism pass freely through the vent, while microorganisms do not.
Order Now
SPECIFICATIONS
Environmentally Preferable
Environmental certification