Avanti™ J-15 Series Benchtop Centrifuges, Beckman Coulter®
Supplier: Beckman Coulter
B99516 C01994 C01995 B99517 C19391 C19392 C19393 C19394 C19397 C19398 C19399 C19400 C19403 C19404 C19405 C19406 C19409 C19410 C19411 C19412 C19415 C19416 C19417 C19418 C19421 C19422 C19423 C19424
CA76121-314EA 20571.37 CAD
CA76121-314 CA76121-334 CA76121-336 CA76121-338 CABKC19391 CABKC19392 CABKC19393 CABKC19394 CABKC19397 CABKC19398 CABKC19399 CABKC19400 CABKC19403 CABKC19404 CABKC19405 CABKC19406 CABKC19409 CABKC19410 CABKC19411 CABKC19412 CABKC19415 CABKC19416 CABKC19417 CABKC19418 CABKC19421 CABKC19422 CABKC19423 CABKC19424
Avanti™ J-15 Series Benchtop Centrifuges, Beckman Coulter®
Centrifuges Benchtop Centrifuges
The Avanti™ J-15 series of benchtop centrifuges (refrigerated or ventilated) leverage the ultra harmonic technology, designed to protect your sample and increase workflow time efficiencies.
  • Ultra harmonic technology
  • Workflow optimization
  • Sample protection
  • Time efficiency
  • Multi-dimension imbalance tolerance
The Avanti™ J-15 series of benchtop centrifuges is part of a new family of life science equipment that provides the exceptional performance.
Refrigerated (Avanti™ J-15R) or ventilated (Avanti™ J-15), these centrifuges use ultra harmonicMore Product Information