Biomek® Pipette Tips for Biomek Liquid Handlers, Beckman Coulter®
Supplier: Beckman Coulter
379501 379502 379503 379504 379505 379506 394263 394622 394627 717251 717252 717253 717254 717255 717256 719011 719012 719013 719222 719223 987925 987935 987936 A21578 A21582 A22287 A22288 A21586 A22287 379503 719222 B01089 B01090 B01091 B01112 B01113 B01114 B01122 B01123 B01124 B01101 B01102 B01103 B85955 B85965 B85981 B85739 B85745 B85753 B85756 B85749 B85760 B85877 B85903 B85945 B85961 B85975 B85936 B85881 B85764 B85884 B85767 B85906 B85926 B85929 B85771 B85888 B85911 B85775 B85915 B85892 B85872 B85917 B85894 B85940 B85959 B85971 B85922 B85899
CABK379501CS 174.4 CAD
CABK379501 CABK379502 CABK379503 CABK379504 CABK379505 CABK379506 CABK394263 CABK394622 CABK394627 CABK717251 CABK717252 CABK717253 CABK717254 CABK717255 CABK717256 CABK719011 CABK719012 CABK719013 CABK719222 CABK719223 CABK987925 CABK987935 CABK987936 CABKA21578 CABKA21582 CABKA22287 CABKA22288 CABKA21586 BKA22287 BK379503 BK719222 CABKB01089 CABKB01090 CABKB01091 CABKB01112 CABKB01113 CABKB01114 CABKB01122 CABKB01123 CABKB01124 CABKB01101 CABKB01102 CABKB01103 CA76111-652 CA76111-654 CA76111-656 CA76111-658 CA76111-660 CA76111-662 CA76111-664 CA76111-666 CA76111-668 CA76111-864 CA76111-882 CA76111-890 CA76111-892 CA76111-894 CA76111-896 CA76112-010 CA76112-012 CA76112-014 CA76112-016 CA76112-018 CA76112-020 CA76112-022 CA76112-024 CA76112-026 CA76112-028 CA76112-030 CA76112-032 CA76112-034 CA76112-036 CA76112-038 CA76112-040 CA76112-042 CA76112-044 CA76112-046 CA76112-048 CA76112-050
Biomek® Pipette Tips for Biomek Liquid Handlers, Beckman Coulter®
Pipette Tips
Biomek® Pipette Tips from Beckman Coulter® are the only tips on the market that are designed, validated, and approved for use on Biomek® Liquid Handlers.
  • Color-coded racks
  • Easy visual checks
Tip definitions, pipetting techniques, and templates are built into Biomek® software. For best in total system performance, use only Beckman Coulter® validated pipette tips on Biomek® workstations.
Products that are designated as Sterile are either sterilized using Gamma radiation or ethylene oxide (EtO) gas in accordance with ANSI/AAMI/150 11135 or ANSI/AAMI/More Product Information