HarvestLine System Liners, Beckman Coulter
Supplier: Beckman Coulter
358299 361690 369264 369256 C15218 C19681 C15216 C15217
CABK358299PK 303.1 CAD
CABK358299 CABK361690 CABK369264 CABK369256 CA76292-070 CA76292-072 CA76292-074 CA76292-076 CA76292-078 CA76292-080 CA76292-082 CA76292-140 CA76292-142 CA76292-818 CA76292-820
HarvestLine System Liners, Beckman Coulter
Bags Media Bags
Faster, safer, more efficient harvesting and now it's all in the bag. Minimize non-value-added tasks e.g. bottle cleaning, autoclaving and scraping. Reduce exposure to potentially harmful biologics.
  • Prevent contamination
  • Reduce repetitive motions
  • Prevent liquid spillage
  • Reduce needed storage space
  • Achieve maximum ample yield
  • Expedite freeze/thaw lysing
Safe, efficient workflows are as critical to study as quality samples and to harvest high-quality samples in fewer steps from prep to cleanup.
Optional sterile bottle liners, optimized speed, safetyMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable