Capping Reagent 80/10/10 THF/Lutidine/Acetic Anhydride (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

BR640-4X2.5L
CABJBR6404X2.5CS 5744.64 CAD
CABJBR6404X2.5
Capping Reagent 80/10/10 THF/Lutidine/Acetic Anhydride (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Capping Reagent

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

Boiling Pt: 11.8 %(V) (main component)
Melting Pt: –108.5 °C (pure substance)
Density: 0.888 g/cm³ (20 °C) (main component)
Flash Pt: –15 °C
Storage Temperature: Ambient
UN: 2924
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR