SPE Columns, Honeywell Burdick
Supplier: Burdick & Jackson
9200 9202 9204 9210 9212 9214 9220 9230 9232 9234 9240 9250 9260 9264 9290 9304
CABJ9200PK 467.25 CAD
CABJ9200 CABJ9202 CABJ9204 CABJ9210 CABJ9212 CABJ9214 CABJ9220 CABJ9230 CABJ9232 CABJ9234 CABJ9240 CABJ9250 CABJ9260 CABJ9264 CABJ9290 CABJ9304
SPE Columns, Honeywell Burdick
Chromatography Columns
Medical-grade polypropylene tubes are packed with carefully selected materials to ensure reproducibility, reliability, and high recovery.
  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality
Columns contain a reservoir, retaining frit, packing material, and containing frit. Basic support material is porous, high surface area silica gel. Also available are Alumina, Florisil®, and ion exchange resins. Columns attach securely to the vacuumMore Product Information