Water with 0,05% (v/v) trifluoroacetic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA

486-4
CABJNP486-20EA 15320.98 CAD
CABJNP486-20 CABJ486-4
Water with 0,05% (v/v) trifluoroacetic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Water with 0,05% (v/v) trifluoroacetic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.9982 g/cm³ (20 °C)
Storage Temperature: Ambient
EINECS: 200-929-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR