2-Propanol:Water solution (70:30)% (v/v), LabReady™, Solvent Blend for HPLC, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

420-4
CABJNP420-20EA 29460.57 CAD
CABJNP420-20 CABJ420-4
2-Propanol:Water solution (70:30)% (v/v), LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
2-Propanol:Water

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 87.7 °C (pure substance)
Melting Pt: –62.1 °C
Density: 0.785 g/cm³ (25 °C) (pure substance)
Storage Temperature: Ambient
MDL Number: MFCD00011674
EINECS: 200-661-7
UN: 1219
ADR: 3 II

Order Now

Specification Test Results

Color (APHA) 10 max.
Isopropyl Alcohol (v/v) 69 - 71 %
Residue 1 mg/L max.
Water (v/v) 29 - 31%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR