1-Propanol ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

322-1L 322-4
CABJ322-1LCS 1027.36 CAD
CABJ322-1L CABJ322-4
1-Propanol ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
1-Propanol

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: H₃CCH₂CH₂OH
Boiling Pt: 97.2 °C
Melting Pt: –126.2 °C
Density: 0.8053 g/cm³ (20 °C)
Flash Pt: 13.7 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,III
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis 99.5 % min.
Refractive Index (20ºC) 1.3841 - 1.3857
Residue 3 mg/L max.
UV Absorbance @ 225 nm 0.5 AU max.
UV Absorbance @ 250 nm 0.05 AU max.
UV Absorbance @ 300 nm 0.005 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 210 nm max.
Water by Karl Fischer Titration 0.05 % max.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR