Potassium hydroxide 1.0 M KOH (1.0 N) volumetric solution, Fluka™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Potash caustic, Caustic potash

319376-2L
CABJ319376-2LEA 139.58 CAD
CABJ319376-2L
Potassium hydroxide 1.0 M KOH (1.0 N) volumetric solution, Fluka™
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 1.05 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

Appearance complying
Coloration complying
Clearness complying
Assay titr. 0.950 - 1.050 mol/l
Traceable to NIST SRM complying
BAM/EMPA reference material complying
BAM/EMPA Certificate Number complying
Carbonate (CO₃) Max. 0.01 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR