Iodine 0.02M in 70/20/10 THF/pyridine/water (v/v) oxidizing reagent, for biosynthesis

Supplier: MilliporeSigma

BI0420-0450 BI0420-4000
CABI0420-0450CS 1185 CAD
CABI0420-0450 CABI0420-4000
Iodine 0.02M in 70/20/10 THF/pyridine/water (v/v) oxidizing reagent, for biosynthesis
Iodine

Oxidizing reagents are involved in the formulations for oligonucleotide synthesis


These are suitable for all kind of biosynthesis processes such as DNA, RNA and peptide synthesis.

Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
MDL Number: MFCD00011355
CAS Number: 7553-56-2
Merck Index: 13,05036

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR