BD Insyte™ Autoguard™ Shielded IV Catheters, BD™
Supplier: BD
381512
CABD381512CS 895.87 CAD
CABD381512 CABD381423
BD Insyte™ Autoguard™ Shielded IV Catheters, BD™
Catheters
These IV catheters feature Autoguard™ technology that instantly retracts the needle with the push of a button
Made from BD Vialon™, a proprietary biomaterial designed to increase ease of penetration and indwelling time while decreasing the risk of infiltration and phlebitis. The specially notched needle uses Instaflash™ Technology to immediately confirm vessel entry. Catheters are color-coded to easily identify gauge. Available in standard or winged format.
Order Now
SPECIFICATIONS
Environmentally Preferable