BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
366664 366668 366408 363080 366430 366480 367001 367587 367671 367814 367820 367871 367874 367884 367886 367922 367960 367962 367977 367985 363083 366643 367812 367815 367841 367856 367861 367863 367878 367880 367899 367921 367925 367983 367986 367988
CABD366664CS 643.7 CAD
CABD366664 CA-BD366668 CA97000-582 CABD363080 CABD366430 CABD366480 CABD367001 CABD367587 CABD367671A CABD367814 CABD367820 CABD367871 CABD367874 CABD367884 CABD367886 CABD367922 CABD367960 CABD367962 CABD367964 CABD367977 CABD367985 CABD367987 CABD363083L CABD366643L CABD367812L CABD367815L CABD367841L CABD367856L CABD367861L CABD367863L CABD367878L CABD367880L CABD367899L CABD367921L CABD367925L CABD367983L CABD367986L CABD367988L CABD367844L CABD368861
BD Vacutainer® Venous Blood Collection Tubes
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Sterile vacuum blood collection BD Vacutainer® tubes are for serum determination
Tubes with a BD Hemogard™ closure incorporate a plastic shield that protects lab personnel from contact with blood on the stopper or around the outer rim of the tube, and from potential splattering when tube is opened. The closure's rubber stopper is recessed inside the plastic shield to isolate blood drops left by the needle.
Order Now
SPECIFICATIONS
Environmentally Preferable