Single-Use Syringe/BD PrecisionGlide™ Needle Combinations, Sterile, BD Medical
Supplier: BD
305062 305620 309570 309571 309575 309580 309581 309582 309623 309624 309626 309632 309633 309642 309643 305060 309577 309645 305064
CABD605062CS 299.01 CAD
CABD605062 CABD305620 CABD309570 CABD309571 CABD309572 CABD309574 CABD309575 CABD309578 CABD309579 CABD309580 CABD309581 CABD309582 CABD309597 CABD309623 CABD309624 CABD309625 CABD309626 CABD309630 CABD309631 CABD309632 CABD309633 CABD309634 CABD309637 CABD309642 CABD309643 CABD309644 BD309630 CABD305060 CABD309569L CABD309577L CABD309640L CABD309645L CA11027-094
Single-Use Syringe/BD PrecisionGlide™ Needle Combinations, Sterile, BD Medical
Syringes Syringes, Medical
Sterile syringe/needle combinations have Slip tip or BD Luer-Lok™ tip.
Thin wall needles have a bigger I.D. without any increase in the O.D. Thin wall needles allow the same flow volume as standard-walled needles one gauge larger.
Needles are made of stainless steel with translucent, color-coded polypropylene hubs.
Caution: Needles are autoclavable after being removed from their easy-peel package. They can be sterilized by ethylene oxide in their easy-peel package.
Order Now
SPECIFICATIONS
Environmentally Preferable