Single-Use Needles, BD Medical
Supplier: BD
305106 305109 305125 305127 305129 305145 305165 305167 305194 305195 305196 305197 305198 305186
CABD305106CS 333.97 CAD
CABD305106 CABD305109 CABD305110 CABD305111 CABD305115 CABD305120 CABD305124 CABD305125 CABD305127 CABD305129 CABD305136 CABD305143 CABD305145 CABD305155 CABD305156 CABD305159 CABD305165 CABD305167 CABD305175 CABD305176 CABD305177 CABD305178 CABD305179 CABD305185 CABD305187 CABD305189 CABD305193 CABD305194 CABD305195 CABD305196 CABD305197 CABD305198 CABD305199 BD305120 BD305186
Single-Use Needles, BD Medical
Syringe Needles
These sterile, nonpyrogenic, nontoxic needles are designed for slip tip, eccentric, or BD Luer-Lok® syringes
The inert hubs are color-coded, with the color repeated on packaging labels.
Ordering information: Packed in sterile strips of five needles.