Bulk Syringes, Nonsterile, BD Medical
Supplier: BD
301025 301027 301028 301029 301030 301031 301034 301035 301073
CABD301025CS 765.88 CAD
CABD301025 CABD301027 CABD301028 CABD301029 CABD301030 CABD301031 CABD301033 CABD301034 CABD301035 CABD301036 CABD301032 CABD301073L
Bulk Syringes, Nonsterile, BD Medical
Syringes Syringes, Medical
Clean, ready-to-use, nonsterile syringes are siliconized and assembled with scale.
  • Syringes can be autoclaved or gas sterilized out of poly bags
  • Before autoclaving, remove plunger rod from barrel
Note: Syringes are supplied without needle. Needles must be purchased separately.
Ordering information: All syringes are bulk packed in double-wrapped poly bags. Sterile tip shield is sold separately.