Ammonia solution 20 - 22%, OmniTrace Ultra™ Trace_metal, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

MQ100

AX1308-7
CAAX1308-7EA 432 CAD
CAAX1308-7
Ammonia solution 20 - 22%, OmniTrace Ultra™ Trace_metal, Supelco®
Ammonia solution

Clear liquid. For low level trace metal analysis. Double-distilled and packaged in ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom conditions. Supplied in specially designed, preleached fluoropolymer resin bottles to guarantee product integrity. Certificate of Analysis supplied with each shipment.

Formula: NH₃
MW: 35.05 g/mol
Density: 0.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672

Order Now

Specification Test Results

Aluminum (Al) 20pg/g max
Antimony (Sb) 10pg/g max
Arsenic (As) 10pg/g max
Assay (as NH3) 20-22%
Barium (Ba) 10pg/g max
Beryllium (Be) 10pg/g max
Bismuth (Bi) 10pg/g max
Cadmium (Cd) 10pg/g max
Calcium (Ca) 10pg/g max
Cerium (Ce) 10pg/g max
Cesium (Cs) 10pg/g max
Chromium (Cr) 10pg/g max
Cobalt (Co) 10pg/g max
Copper (Cu) 10pg/g max
Dysprosium (Dy) 10pg/g max
Erbium (Er) 10pg/g max
Europium (Eu) 10pg/g max
Gadolinium (Gd) 10pg/g max
Gallium (Ga) 10pg/g max
Germanium (Ge) 10pg/g max
Gold (Au) 10pg/g max
Holmium (Ho) 10pg/g max
Indium (In) 10pg/g max
Iron (Fe) 10pg/g max
Lanthanum (La) 10pg/g max
Lead (Pb) 10pg/g max
Lithium (Li) 10pg/g max
Lutetium (Lu) 10pg/g max
Magnesium (Mg) 10pg/g max
Manganese (Mn) 10pg/g max
Mercury (Hg) 200pg/g max
Molybdenum (Mo) 10pg/g max
Neodymium (Nd) 10pg/g max
Nickel (Ni) 10pg/g max
Niobium (Nb) 10pg/g max
Potassium (K) 10pg/g max
Praseodymium (Pr) 10pg/g max
Rhodium (Rh) 10pg/g max
Rubidium (Rb) 10pg/g max
Samarium (Sm) 10pg/g max
Scandium (Sc) 10pg/g max
Silver (Ag) 10pg/g max
Sodium (N/A) 20pg/g max
Strontium (Sr) 10pg/g max
Tellurium (Te) 10pg/g max
Terbium (Tb) 10pg/g max
Thallium (Tl) 10pg/g max
Thorium (Th) 10pg/g max
Thulium (Tm) 10pg/g max
Tin (Sn) 10pg/g max
Titanium (Ti) 10pg/g max
Tungsten (W) 10pg/g max
Uranium (U) 10pg/g max
VaN/Adium (V) 10pg/g max
Ytterbium (Yb) 10pg/g max
Yttrium (Y) 10pg/g max
Zinc (Zn) 10pg/g max
Zirconium (Zr) 10pg/g max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR