Ammonium hydrogen difluoride ≥96.0%, Supelco®

Supplier: MilliporeSigma
Danger

AX1235-5 AX1235-1
CAAX1235-5EA 893 CAD
CAAX1235-5 CAAX1235-1
Ammonium hydrogen difluoride ≥96.0%, Supelco®
Ammonium hydrogen difluoride

Practical

Formula: NH₄HF₂
Boiling Pt: 238 °C (1013 hPa) (dec.)
Melting Pt: 126 °C
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS Number: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Merck Index: 13,00498

Order Now

Specification Test Results

Assay 96% min
Color White (typical)
Form Flakes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR