Acetone 99,5 GC, corrected for water content HPLC, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

AX0115-20 AX0115-6 AX0115-NP20 AX0115-1 AX0115P-1 AX0115P-6
CAAX0115-20EA 5733 CAD
CAAX0115-20 CAAX0115-38 CAAX0115-39 CAAX0115-40 CAAX0115-6 CAAX0115-NP20 CAAX0115-1 CAAX0115P-1 CAAX0115P-6
Acetone 99,5 GC, corrected for water content HPLC, Supelco®
Acetone

HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs.

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC, corrected for water content) 99.5% min
Aldehyde (as HCHO) 0.002% max
Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Liquid
Infrared spectrum Conforms to standard
Isopropyl alcohol 0.05% max
Methanol 0.05% max
Residue after evaporation 5ppm max
Solubility in water To pass test
Substances reducing permanganate To pass test
Titrable acid 0.0003meq/g max
Titrable base 0.0006meq/g max
UV Abs. at 330 nm 1.00AU max
UV Abs. at 340 nm 0.10AU max
UV Abs. at 350 nm 0.02AU max
UV Abs. at 400 nm 0.01AU max
UV Cut-off 330nm max
Water (H2O) 0.5% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR