PL Multisolvent, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
PL1515-3323
CAAGPL15153323EA 2451.25 CAD
CAAGPL15153323
PL Multisolvent, Agilent Technologies
Chromatography Columns
PL Multisolvent GPC columns use polymer-coated silica to operate in a wide range of solvents.
  • The PL Multisolvent has the efficiency and robustness of silica with a special coating to eliminate ‘sticking’
  • Silica backbone provides the high strength necessary for high-efficiency particles and the rigid structure that eliminates pore swelling and collapse with solvent switching
  • High-resolution separation of polymers up to 100,000 Da, as well as additives, oligomers, and plasticizers
The PL Multisolvent family uses a revolutionary process to coat high-More Product Information