Source Lamps for UV-Vis & UV-Vis-NIR, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
110713990 2140-0605 5610013900 5610021700 G9820-80000 G9820-60000
CA110713990EA 1078.8 CAD
CA110713990 CA76514-586 CA5610013900 CA5610021700 CAG9820-80000 CAAGG982060000
Source Lamps for UV-Vis & UV-Vis-NIR, Agilent Technologies
Lights Deuterium Lamps
Deuterium and Visible source lamps provide good stability and intensity over the wavelength range.
  • Designed to provide optimal intensity, sensitivity, and stability
  • Built to meet highest-quality standards and strictest safety regulation
  • Rigorously tested for best lamp-to-lamp consistency
  • Robust and long-lasting to lower your cost of ownership
  • Extensively tested to ensure they meet the tightest specifications and quality standards
  • Recommended for use with Agilent UV-Vis and UV-Vis-NIR instruments
Deuterium and Visible source lamps provide goodMore Product Information