Sample Loops for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G8010-60305 G8010-60338 G8010-60339 G8010-60341 G8010-60340 G8010-60342 G8010-60343 G8010-60344
CAAGG801060305EA 309.49 CAD
CAAGG801060305 CAAGG801060338 CAAGG801060339 CAAGG801060341 CAAG9910143300 CAAG9910143400 CAAGG801060340 CAAGG801060342 CAAGG801060343 CAAGG801060344
Sample Loops for ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Sample Loops
Sample loops in various sizes for ICP-OES SVS1/2/2+ and AVS 4/6/7 systems.
  • Wide variety of sample loop sizes available to meet your application needs
  • Sample loops are labeled to easily identify loop size
  • With the AVS 6/7 with bubble injection a 1 ml sample loop provides 47 seconds of sample measurement
  • Smaller loops can be used to further reduce uptake delay as well as reduce rinse times
Agilent sample loops for SVS 1/2/2+ and AVS 4/6/7 are available in a wide variety of sizes to meet specific application needs. ICP-OES Sample Loops are available in 0.More Product Information