Miscellaneous Supplies for MP-AES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G8000-64112
CAAGG800064112EA 182.65 CAD
CAAGG800064112
Miscellaneous Supplies for MP-AES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Supplies for MP-AES include Pre-optic window and disposable Onboard Argon bottle for plasma ignition.
  • Miscellaneous supplies can help you to maintain the superior performance of the MP-AES
  • Easily order for your planned maintenance and ensuring your supplies cabinet is properly provisioned
Agilent can help you to maintain the superior performance of the MP-AES with all the supplies and accessories you need. Miscellaneous supplies include user replaceable Pre-optic window for MP-AES, and disposable Onboard Argon bottle for plasma ignition (not available in all countries).