CrossLab Needle and Needle Seats for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G1313-87102 G1329-87101 G1367-87102 G1377-87201 G2260-87101 G1367-87101 G1367-87201 G1329-87012 G1329-87017 G1367-87012 G1367-87017 G4226-87012 G4226-87201 G4267-87012 G4267-87201 G7129-87012 G7129-87017 G1367-87202 G2260-87201 G5667-81008 G5667-87200 G7129-87117 G7167-68711 5188-8052 G2258-68710 G2258-87102 G4226-87211 G4267-87020 G4267-87210 G7129-87112 G7157-87101 G7157-87201 G1329-87103 G1377-87000 G1377-87001 G1377-87002 G4226-87030 G7137-60075 G7137-87012 G7137-87201 G7167-87308
CAG1313-87102EA 436.01 CAD
CAG1313-87102 CAG1329-87101 CAG1367-87102 CAG1377-87201 CAG2260-87101 CA76508-254 CA76515-228 CAG1329-87012 CAG1329-87017 CAG1367-87012 CAG1367-87017 CAG4226-87012 CAG4226-87201 CAG4267-87012 CAG4267-87201 CAG7129-87012 CAG7129-87017 CAG1367-87202 CAG2260-87201 CAG5667-81008 CAG5667-87200 CAG7129-87117 CAG7167-68711 CAAG5188-8052 CAAGG225868710 CAAGG225887102 CAAGG422687211 CAAGG426787020 CAAGG426787210 CAAGG712987112 CAAGG715787101 CAAGG715787201 CAAGG132987103 CAAGG137787000 CAAGG137787001 CAAGG137787002 CAAGG422687030 CAAGG713760075 CAAGG713787012 CAAGG713787201 CAAGG716787308
CrossLab Needle and Needle Seats for HPLC, Agilent Technologies
Syringe Needles
A comprehensive range of needles and needle seats for HPLC samplers.
  • The needle should be replaced when it becomes bent, burred, or blunt, or when it is leaking or plugged
  • Suspect a leak if you notice a trail of buffer crystals on the needle seat
  • If the sample contains particles, the needle seat can become blocked, as this is the first restriction that the sample experiences
  • If this occurs, try backflushing the needle seat capillary
Agilent provides a comprehensive range of needles and needle seats to equip all different HPLC samplers. SpecificMore Product Information