Hydride Generation Supplies for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3138-65128
CAAGG313865128PK 335.15 CAD
CAAGG313865128
Hydride Generation Supplies for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
ISIS Hydride Generation Accessories for analysis of As, Sb, Se, Sn and Hg with ICP-MS systems.
  • ISIS hydride generation accessories are specially designed for Agilent instruments to ensure high quality results
  • Flexible selection of accessories for every instrument configuration
  • Convenient kits and accessories are simple to order and make user maintenance easy to schedule and execute
  • Hydride generation offers the best sensitivity for As, Sb, Se, Sn and Hg with ICP-MS, which are all hydride forming elements
Agilent manufactures a range of high qualityMore Product Information