CrossLab Instrument Supplies for GC/MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
0515-1446 05971-20134 05971-60140 05990-60084 19231-20950 19231-20960 19231-20970 G1072-20008 G1099-20130 G1099-20136 G1999-20021 G2589-20043 G2589-20100 5061-5869 0100-2138 05971-20143 05971-80103 5062-3506 G1099-20132 G1099-20133 G2589-20044 G2589-20045 G3170-20126 G3170-20530 392017401 393010918 G2571-67001 G3163-20530 G3870-20444 G3870-20445 G3870-20448 G3870-20449 G3870-20530 G7002-20074 G7002-80103 393031501 393059191 5190-9071 G1999-20432 G1999-20446 G1999-60412 G3170-20540 G3170-60172 G3170-80002 G3440-20022 G3870-20440 G3870-67171 G7000-20026 G7002-20064 G7002-20083 G7002-20109 G7002-67057 392043700 393175101 393847701 5062-3507 G1099-80001 G1999-20443 G2589-20101 G3169-60170 G3170-20430 G3170-60171 G3170-60416
CA0515-1446EA 6.78 CAD
CA0515-1446 CA05971-20134 CA05971-60140 CA05990-60084 CA19231-20950 CA19231-20960 CA19231-20970 CAG1072-20008 CAG1099-20130 CAG1099-20136 CAG1999-20021 CAG2589-20043 CAG2589-20100 CA5061-5869 CA0100-2138 CA76514-518 CA76514-520 CA76514-658 CA76515-168 CA76515-170 CA76515-260 CAG2589-20045 CAG3170-20126 CAG3170-20530 CA392017401 CA393010918 CAG2571-67001 CAG3163-20530 CAG3870-20444 CAG3870-20445 CAG3870-20448 CAG3870-20449 CAG3870-20530 CAG7002-20074 CAG7002-80103 CAAG393031501 CAAG393059191 CAAG5190-9071 CAAGG199920432 CAAGG199920446 CAAGG199960412 CAAGG317020540 CAAGG317060172 CAAGG317080002 CAAGG344020022 CAAGG387020440 CAAGG387067171 CAAGG700020026 CAAGG700220064 CAAGG700220083 CAAGG700220109 CAAGG700267057 CAAG392043700 CAAG393011391 CAAG393175101 CAAG393847701 CAAG5062-3507 CAAGG109980001 CAAGG199920443 CAAGG258920101 CAAGG316960170 CAAGG317020430 CAAGG317060171 CAAGG317060416
CrossLab Instrument Supplies for GC/MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Maintain your GC/MS system with Agilent supplies for optimal results.
  • A comprehensive range of supplies to help you maintain GC/MS performance
  • Ion source filaments to facilitate regular replacement
  • Electron multipliers for various Agilent MS instruments
Your mass spectrometer is a sensitive, specialized device that delivers a higher level of functionality compared to other GC detectors. To continue to achieve optimal results, diligent system maintenance is required. The Agilent range of GC/MS instrument supplies include: MS filaments, electronMore Product Information